Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution
Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution
Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution
Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution
Thermo Scientific Chemicals
Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution
CAS: 16940-66-2 | BH4Na | 37.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic bottle
Catalog number FSA389930025
Price (MYR)
1,172.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,172.00
BOT
Add to cart
Chemical Identifiers
CAS16940-66-2, 1310-73-2, 67-56-1, 7732-18-5
Specifications
Boiling Point130.0°C to 135.0°C
Chemical Name or MaterialSodium borohydride
Name Note12% solution in 40% aq. sodium hydroxide solution
CAS16940-66-2
ColorColorless
View more
Reduction reagent
RUO – Research Use Only