1-Propanol, for HPLC
1-Propanol, for HPLC
1-Propanol, for HPLC
1-Propanol, for HPLC
Thermo Scientific Chemicals
1-Propanol, for HPLC
CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA389600025
Price (MYR)
1,022.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,022.00
EA
Add to cart
Specifications
Chemical Name or Material1-Propanol
Melting Point-127.0°C
Absorbance0.05 max. (1 cm cell vs HPLC−grade water) at 250nm, 0.3 max. (1 cm cell vs HPLC−grade water) at 220nm, 0.7 max. (1 cm cell vs HPLC−grade water) at 210nm
CAS71-23-8
ColorColorless
View more
For HPLC
RUO – Research Use Only