Water, for analysis
Water, for analysis
Water, for analysis
Thermo Scientific Chemicals
Water, for analysis
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number FSA389400100
Price (MYR)
949.00
EA
Add to cart
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
949.00
EA
Add to cart
Specifications
Absorbance0.01 max. (water, 1cm, 254nm)
Chemical Name or MaterialWater
pH5 to 8
CAS7732-18-5
ColorColorless
View more
Biological buffer
RUO – Research Use Only