Water, for HPLC
Water, for HPLC
Water, for HPLC
Thermo Scientific Chemicals
Water, for HPLC
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA389390025
Price (MYR)
427.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
427.00
BOT
Add to cart
Specifications
Absorbance0.004 max. at 254nm, 0.009 max. at 210nm, 0.05 max. at 200nm
Chemical Name or MaterialWater
CAS7732-18-5
ColorColorless
Health Hazard 1
View more
For HPLC
RUO – Research Use Only