Sodium cyclamate, 99%
Sodium cyclamate, 99%
Sodium cyclamate, 99%
Sodium cyclamate, 99%
Thermo Scientific Chemicals
Sodium cyclamate, 99%
CAS: 139-05-9 | C6H12NNaO3S | 201.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA389282500
Price (MYR)
417.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
417.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium cyclamate
CAS139-05-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.