Calcium chloride hexahydrate, 97+%, extra pure
Calcium chloride hexahydrate, 97+%, extra pure
Calcium chloride hexahydrate, 97+%, extra pure
Calcium chloride hexahydrate, 97+%, extra pure
Thermo Scientific Chemicals
Calcium chloride hexahydrate, 97+%, extra pure
CAS: 7774-34-7 | CaCl2·6H2O | 219.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA389250010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium chloride hexahydrate
Melting Point30.0°C
CAS7774-34-7
ColorColorless
Health Hazard 1GHS Signal Word: Warning