n-Heptane, 99.5%, for analysis
n-Heptane, 99.5%, for analysis
n-Heptane, 99.5%, for analysis
n-Heptane, 99.5%, for analysis
Thermo Scientific Chemicals
n-Heptane, 99.5%, for analysis
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number FSA389170025
Price (MYR)
992.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
992.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Heptane
Melting Point-91.0°C
CAS142-82-5
CAS Min %99.4
Health Hazard 1Danger
View more
For analysis
RUO – Research Use Only