Phosphoric acid, 98+%, pure
Phosphoric acid, 98+%, pure
Phosphoric acid, 98+%, pure
Phosphoric acid, 98+%, pure
Thermo Scientific Chemicals
Phosphoric acid, 98+%, pure
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA389025000
Price (MYR)
522.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
522.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhosphoric acid
ColorWhite
CAS7664-38-2
Melting Point42.0°C
Health Hazard 1Danger