Zinc(II) acetylacetonate, ca. 25% Zn
Zinc(II) acetylacetonate, ca. 25% Zn
Zinc(II) acetylacetonate, ca. 25% Zn
Zinc(II) acetylacetonate, ca. 25% Zn
Thermo Scientific Chemicals
Zinc(II) acetylacetonate, ca. 25% Zn
CAS: 14024-63-6 | C10H14O4Zn | 263.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA388770050
Price (MYR)
285.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
285.00
BOT
Add to cart
Specifications
Boiling Point129.0°C to 131.0°C (10.0mmHg)
Chemical Name or MaterialZinc(II) acetylacetonate
Melting Point124.0°C to 126.0°C
CAS14024-63-6
ColorWhite to Ivory