Ammonium trifluoromethanesulfonate, 99%
Ammonium trifluoromethanesulfonate, 99%
Ammonium trifluoromethanesulfonate, 99%
Ammonium trifluoromethanesulfonate, 99%
Thermo Scientific Chemicals
Ammonium trifluoromethanesulfonate, 99%
CAS: 38542-94-8 | CH4F3NO3S | 167.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA388730500
Price (MYR)
2,720.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,720.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium trifluoromethanesulfonate
Melting Point225°C
CAS38542-94-8
ColorWhite
Health Hazard 1GHS Signal Word: Warning