Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA388381000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS124-40-3, 109-99-9
Specifications
Chemical Name or MaterialDimethylamine
ColorYellow
Name Note2M solution in THF
CAS124-40-3
Health Hazard 1Danger