Trimethylamine, 1M solution in THF, AcroSeal™
Trimethylamine, 1M solution in THF, AcroSeal™
Trimethylamine, 1M solution in THF, AcroSeal™
Trimethylamine, 1M solution in THF, AcroSeal™
Trimethylamine, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals
Trimethylamine, 1M solution in THF, AcroSeal™
CAS: 75-50-3 | C3H9N | 59.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA388371000
Price (MYR)
666.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
666.00
BOT
Add to cart
Chemical Identifiers
CAS75-50-3, 109-99-9
Specifications
Chemical Name or MaterialTrimethylamine
ColorColorless to Yellow
Name Note1M solution in THF
CAS75-50-3
Health Hazard 1Danger