Gold(I) chloride, 99+%
Gold(I) chloride, 99+%
Gold(I) chloride, 99+%
Gold(I) chloride, 99+%
Thermo Scientific Chemicals
Gold(I) chloride, 99+%
CAS: 10294-29-8 | AuCl | 232.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA388330050
Price (MYR)
9,289.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
9,289.00
EA
Add to cart
Specifications
Chemical Name or MaterialGold(I) chloride
Melting Point289.0°C
CAS10294-29-8
ColorYellow to Green
Health Hazard 1GHS Signal Word: Danger