5-chloro-2-pentanone, 90%
5-chloro-2-pentanone, 90%
5-chloro-2-pentanone, 90%
5-chloro-2-pentanone, 90%
Thermo Scientific Chemicals
5-chloro-2-pentanone, 90%
CAS: 5891-21-4 | C5H9ClO | 120.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA388291000
Price (MYR)
1,318.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,318.00
EA
Add to cart
Specifications
Chemical Name or Material5-Chloro-2-pentanone
CAS5891-21-4
CAS Min %88.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0