Bis(trifluoromethanesulfonyl)imide, 99%
Bis(trifluoromethanesulfonyl)imide, 99%
Bis(trifluoromethanesulfonyl)imide, 99%
Bis(trifluoromethanesulfonyl)imide, 99%
Thermo Scientific Chemicals
Bis(trifluoromethanesulfonyl)imide, 99%
CAS: 82113-65-3 | C2HF6NO4S2 | 281.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA387600250
Price (MYR)
3,229.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,229.00
EA
Add to cart
Specifications
Chemical Name or MaterialBis(trifluoromethanesulfonyl)imide
CAS82113-65-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.
Health Hazard 3GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Coupling reagent
RUO – Research Use Only