Nickel(II) chloride, 98%
Nickel(II) chloride, 98%
Nickel(II) chloride, 98%
Nickel(II) chloride, 98%
Thermo Scientific Chemicals
Nickel(II) chloride, 98%
Nickel(II) chloride, 98%, Cl2Ni, CAS Number-7718-54-9, 250g, 97.5% min. (Argentometry), 100.0, CHEBI:34887, Brown to Orange, 231-743-, 129.6, GHS Signal Word: Danger | CAS: 7718-54-9 | Cl2Ni | 129.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA387322500
Price (MYR)
1,102.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,102.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNickel(II) chloride
Melting Point1001.0°C
CAS7718-54-9
ColorBrown to Orange
CAS Min %97.5