4,4'-Biphenyldicarboxylic acid, 98%
4,4'-Biphenyldicarboxylic acid, 98%
4,4'-Biphenyldicarboxylic acid, 98%
4,4'-Biphenyldicarboxylic acid, 98%
Thermo Scientific Chemicals
4,4'-Biphenyldicarboxylic acid, 98%
CAS: 787-70-2 | C14H10O4 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA387300250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4, 4'-Biphenyldicarboxylic acid
CAS787-70-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0