3,5-Di-tert-butylsalicylic acid, 99%
3,5-Di-tert-butylsalicylic acid, 99%
3,5-Di-tert-butylsalicylic acid, 99%
3,5-Di-tert-butylsalicylic acid, 99%
Thermo Scientific Chemicals
3,5-Di-tert-butylsalicylic acid, 99%
CAS: 19715-19-6 | C15H22O3 | 250.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA386860050
Price (MYR)
276.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
276.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 5-Di-tert-butylsalicylic acid
CAS19715-19-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0