Pyridine, 99+%, for spectroscopy
Pyridine, 99+%, for spectroscopy
Pyridine, 99+%, for spectroscopy
Pyridine, 99+%, for spectroscopy
Thermo Scientific Chemicals
Pyridine, 99+%, for spectroscopy
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass Bottle
Catalog number FSA386560250
Price (MYR)
335.00
ML
Add to cart
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
335.00
ML
Add to cart
Specifications
Chemical Name or MaterialPyridine
Melting Point-42.0°C
Name Note99+%
Absorbance0.022 max. at 345nm, 0.097 max. at 340nm, 0.301 max. at 335nm, 1 max. at 310nm
CAS110-86-1
View more
For UV/VIS spectroscopy
RUO – Research Use Only