L(+)-Glutamine, specified according to the requirements of USP
CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol
Have Questions?
Change view
Quantity:
250 g
50 g
1 kg
Catalog number FSA386032500
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL(+)-Glutamine
Melting Point185.0°C
CAS56-85-9
Molecular FormulaC5H10N2O3
Linear FormulaH2NCOCH2CH2CH(NH2)CO2H