Citric acid monohydrate, 99+%, ACS reagent
Citric acid monohydrate, 99+%, ACS reagent
Citric acid monohydrate, 99+%, ACS reagent
Citric acid monohydrate, 99+%, ACS reagent
Thermo Scientific Chemicals
Citric acid monohydrate, 99+%, ACS reagent
CAS: 5949-29-1 | C6H8O7·H2O | 210.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA385850010
Price (MYR)
439.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
439.00
EA
Add to cart
Specifications
Chemical Name or MaterialCitric acid monohydrate
CAS5949-29-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact len