Sodium dichromate dihydrate, ACS reagent, 99.5-100.5%
Sodium dichromate dihydrate, ACS reagent, 99.5-100.5%
Sodium dichromate dihydrate, ACS reagent, 99.5-100.5%
Sodium dichromate dihydrate, ACS reagent, 99.5-100.5%
Thermo Scientific Chemicals
Sodium dichromate dihydrate, ACS reagent, 99.5-100.5%
CAS: 7789-12-0 | Cr2Na2O7·2H2O | 298 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA385631000
Price (MYR)
313.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
313.00
EA
Add to cart
Specifications
Boiling Point400.0°C
Chemical Name or MaterialSodium dichromate dihydrate
Melting Point357.0°C
CAS7789-12-0
Health Hazard 1GHS Signal Word: Danger