Methyl nonanoate, 95%
Methyl nonanoate, 95%
Methyl nonanoate, 95%
Methyl nonanoate, 95%
Thermo Scientific Chemicals
Methyl nonanoate, 95%
CAS: 1731-84-6 | C10H20O2 | 172.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA385560250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl nonanoate
CAS1731-84-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
Health Hazard 3GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.