Propyl propionate, 98+%
Propyl propionate, 98+%
Propyl propionate, 98+%
Propyl propionate, 98+%
Thermo Scientific Chemicals
Propyl propionate, 98+%
CAS: 106-36-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA385522500
Price (MYR)
356.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
356.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPropyl propionate
CAS106-36-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if inhaled.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a