Valinomycin, 90%
Valinomycin, 90%
Valinomycin, 90%
Thermo Scientific Chemicals
Valinomycin, 90%
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
Packaging:
Glass Bottle
Catalog number FSA385120500
Price (MYR)
1,719.00
EA
Add to cart
Quantity:
50 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,719.00
EA
Add to cart
Specifications
Chemical Name or MaterialValinomycin
TypeValinomycin
Physical FormCrystalline Powder
CAS2001-95-8
Health Hazard 1GHS Signal Word: Danger