1-Octadecanesulfonic acid, sodium salt, 90%
1-Octadecanesulfonic acid, sodium salt, 90%
1-Octadecanesulfonic acid, sodium salt, 90%
1-Octadecanesulfonic acid, sodium salt, 90%
Thermo Scientific Chemicals
1-Octadecanesulfonic acid, sodium salt, 90%
CAS: 13893-34-0 | C18H37NaO3S | 356.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA384910100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Octadecanesulfonic acid, sodium salt
CAS13893-34-0
CAS Min %88.0
CAS Max %100.0
Melting Point163°C to 165°C