Ammonium thioglycolate, 70 wt.% solution in water
Ammonium thioglycolate, 70 wt.% solution in water
Ammonium thioglycolate, 70 wt.% solution in water
Ammonium thioglycolate, 70 wt.% solution in water
Thermo Scientific Chemicals
Ammonium thioglycolate, 70 wt.% solution in water
Ammonium thioglycolate, 68.0 to 72.0%, C2H7NO2S, CAS Number-5421-46-5, 7732-18-5 | CAS: 5421-46-5 | C2H7NO2S | 109.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number FSA384682500
Price (MYR)
288.00
BOT
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
288.00
BOT
Add to cart
Chemical Identifiers
CAS5421-46-5, 7732-18-5
Specifications
Chemical Name or MaterialAmmonium thioglycolate
ColorColorless
Name Note70 wt.% solution in water
CAS5421-46-5
Boiling Point115.0°C