3,4-difluorobenzyl Chloride, 98%
3,4-difluorobenzyl Chloride, 98%
3,4-difluorobenzyl Chloride, 98%
3,4-difluorobenzyl Chloride, 98%
Thermo Scientific Chemicals
3,4-difluorobenzyl Chloride, 98%
CAS: 698-80-6 | C7H5ClF2 | 162.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA384420250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 4-Difluorobenzyl chloride
CAS698-80-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0