Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Thermo Scientific Chemicals
Isopropanol, 99.5%, for HPLC
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA383910010
Price (MYR)
623.00
BOT
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
623.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
CAS67-63-0
Density0.7850g/mL
Boiling Point81.0°C to 83.0°C