n-Pentane, for analysis
n-Pentane, for analysis
n-Pentane, for analysis
n-Pentane, for analysis
Thermo Scientific Chemicals
n-Pentane, for analysis
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA383900025
Price (MYR)
772.00
LT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
772.00
LT
Add to cart
Specifications
Chemical Name or Materialn-Pentane
Melting Point-130.0°C
CAS109-66-0
Health Hazard 1Danger
Health Hazard 2GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
View more
For analysis
RUO – Research Use Only