Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use
Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use
Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use
Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use
Thermo Scientific Chemicals
Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use
CAS: 1330-43-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number FSA383875000
Price (MYR)
312.00
ML
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
312.00
ML
Add to cart
Chemical Identifiers
CAS1330-43-4, 7732-18-5, 7647-14-5
Specifications
Chemical Name or MaterialBuffer solution pH 9.2
Name NoteBorate buffer, traceable to NIST, ready to use
CAS1330-43-4
CAS7732-18-5
Density1.0000g/mL