Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Thermo Scientific Chemicals
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
CAS: 7558-79-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number FSA383855000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS7558-79-4, 7732-18-5, 7487-94-7, 7778-77-0, 7647-14-5
Specifications
Chemical Name or MaterialBuffer solution pH 7
Name NotePhosphate buffer, traceable to NIST, ready to use
CAS7558-79-4
CAS7732-18-5
Density1.0000g/mL