Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
Thermo Scientific Chemicals
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
CAS: 877-24-7 | C8H5KO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Catalog number FSA383835000
Price (MYR)
169.00
ML
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
169.00
ML
Add to cart
Chemical Identifiers
CAS877-24-7, 7732-18-5, 7487-94-7
Specifications
Chemical Name or MaterialBuffer solution pH 4
CAS877-24-7
CAS7732-18-5
CAS7487-94-7
Concentration1.0% (Potassium hydrogen phthalate)