Isohexane, for HPLC, contains <5% n-Hexane
Isohexane, for HPLC, contains &lt;5% n-Hexane
Isohexane, for HPLC, contains &lt;5% n-Hexane
Isohexane, for HPLC, contains &lt;5% n-Hexane
Thermo Scientific Chemicals
Isohexane, for HPLC, contains <5% n-Hexane
CAS: 73513-42-5 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA383810010
Price (MYR)
1,108.00
LT
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,108.00
LT
Add to cart
Specifications
Chemical Name or MaterialIsohexane
CAS73513-42-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a