Chloroform, 99.8%, for analysis, stabilized with amylene
Chloroform, 99.8%, for analysis, stabilized with amylene
Chloroform, 99.8%, for analysis, stabilized with amylene
Chloroform, 99.8%, for analysis, stabilized with amylene
Thermo Scientific Chemicals
Chloroform, 99.8%, for analysis, stabilized with amylene
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number FSA383770025
Price (MYR)
735.00
LT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
735.00
LT
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteStabilized with amylene
CAS67-66-3
Health Hazard 1Warning
View more
For analysis
RUO – Research Use Only