Bis(2-diphenylphosphinophenyl)ether, 99%
Bis(2-diphenylphosphinophenyl)ether, 99%
Bis(2-diphenylphosphinophenyl)ether, 99%
Bis(2-diphenylphosphinophenyl)ether, 99%
Thermo Scientific Chemicals
Bis(2-diphenylphosphinophenyl)ether, 99%
CAS: 166330-10-5 | C36H28OP2 | 538.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA383370250
Price (MYR)
1,444.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,444.00
EA
Add to cart
Specifications
Chemical Name or MaterialBis(2-diphenylphosphinophenyl)ether
CAS166330-10-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0