Samarium(III) trifluoromethanesulfonate, 98%
Samarium(III) trifluoromethanesulfonate, 98%
Samarium(III) trifluoromethanesulfonate, 98%
Samarium(III) trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals
Samarium(III) trifluoromethanesulfonate, 98%
CAS: 52093-28-4 | C3F9O9S3Sm | 597.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Plastic bottle
Catalog number FSA383170100
Price (MYR)
880.00
EA
Add to cart
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
880.00
EA
Add to cart
Specifications
Chemical Name or MaterialSamarium(III) trifluoromethanesulfonate
CAS52093-28-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: