Dibutyltin dilaurate, 94%
Dibutyltin dilaurate, 94%
Dibutyltin dilaurate, 94%
Dibutyltin dilaurate, 94%
Thermo Scientific Chemicals
Dibutyltin dilaurate, 94%
CAS: 77-58-7 | C32H64O4Sn | 631.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3826900505 gPlastic bottle
Catalog number FSA382690050
Price (MYR)
281.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
281.00
BOT
Add to cart
Specifications
Boiling Point205.0°C (12.0 mmHg)
Chemical Name or MaterialDibutyltin dilaurate
Melting Point16.0°C to 18.0°C
CAS77-58-7
ColorYellow