Cellulose microcrystalline, extra pure, average particle size 90 μm
Cellulose microcrystalline, extra pure, average particle size 90 μm
Cellulose microcrystalline, extra pure, average particle size 90 μm
Cellulose microcrystalline, extra pure, average particle size 90 μm
Thermo Scientific Chemicals
Cellulose microcrystalline, extra pure, average particle size 90 μm
CAS: 9004-34-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA382312500
Price (MYR)
353.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
353.00
EA
Add to cart
Specifications
Chemical Name or MaterialCellulose microcrystalline
ColorWhite
pH5 to 7
CAS9004-34-6
Physical FormFine Crystalline Powder or Granular Powder
View more
Filter aid
RUO – Research Use Only