4-(Aminomethyl)pyridine, 98%
4-(Aminomethyl)pyridine, 98%
4-(Aminomethyl)pyridine, 98%
4-(Aminomethyl)pyridine, 98%
Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 98%
CAS: 3731-53-1 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSA382081000
Price (MYR)
922.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
922.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Aminomethyl)pyridine
CAS3731-53-1
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
Melting Point-8.0°C