Trimethylsulfonium hydroxide, 0.25M solution in methanol
Trimethylsulfonium hydroxide, 0.25M solution in methanol
Trimethylsulfonium hydroxide, 0.25M solution in methanol
Trimethylsulfonium hydroxide, 0.25M solution in methanol
Thermo Scientific Chemicals
Trimethylsulfonium hydroxide, 0.25M solution in methanol
CAS: 17287-03-5 | C3H9IS | 204.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Catalog number FSA381360100
Price (MYR)
1,136.00
BOT
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (MYR)
1,136.00
BOT
Add to cart
Chemical Identifiers
CAS17287-03-5, 67-56-1
Specifications
Chemical Name or MaterialTrimethylsulfonium hydroxide
ColorColorless
Name Note0.25M solution in methanol
CAS17287-03-5
Health Hazard 1GHS Signal Word: Danger