Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™
Thermo Scientific Chemicals
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™
CAS: 96-10-6 | C4H10AlCl | 120.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA381151000
Price (MYR)
580.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
580.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiethylaluminium chloride
CAS96-10-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damag
CAS108-88-3
View more
Lewis acid
RUO – Research Use Only