Lithium Bis(trifluoromethanesulfonimide), 99%
Lithium Bis(trifluoromethanesulfonimide), 99%
Lithium Bis(trifluoromethanesulfonimide), 99%
Lithium Bis(trifluoromethanesulfonimide), 99%
Thermo Scientific Chemicals
Lithium Bis(trifluoromethanesulfonimide), 99%
CAS: 90076-65-6 | C2F6LiNO4S2 | 287.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA381030500
Price (MYR)
2,456.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,456.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLithium bis(trifluoromethanesulfonimide)
CAS90076-65-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.<
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins