Potassium hydroxide, pure, 8N solution in water
Potassium hydroxide, pure, 8N solution in water
Potassium hydroxide, pure, 8N solution in water
Potassium hydroxide, pure, 8N solution in water
Thermo Scientific Chemicals
Potassium hydroxide, pure, 8N solution in water
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA380625000500 mLPlastic bottle
Catalog number FSA380625000
Price (MYR)
288.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
288.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydroxide
CAS1310-58-3
ColorColorless
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May be corrosive to metals.