4-Ethynylanisole, 97%
4-Ethynylanisole, 97%
4-Ethynylanisole, 97%
4-Ethynylanisole, 97%
Thermo Scientific Chemicals
4-Ethynylanisole, 97%
CAS: 768-60-5 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA379920050
Price (MYR)
1,160.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,160.00
EA
Add to cart
Specifications
Chemical Name or Material4-Ethynylanisole
CAS768-60-5
CAS Min %96.0
CAS Max %100.0
Health Hazard 2GHS H Statement: