Lithium hydroxide, 98%, pure, anhydrous
Lithium hydroxide, 98%, pure, anhydrous
Lithium hydroxide, 98%, pure, anhydrous
Lithium hydroxide, 98%, pure, anhydrous
Thermo Scientific Chemicals
Lithium hydroxide, 98%, pure, anhydrous
CAS: 1310-65-2 | HLiO | 23.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA379185000
Price (MYR)
2,488.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,488.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLithium hydroxide
Melting Point462.0°C
CAS1310-65-2
ColorWhite to Yellow
Health Hazard 1GHS Signal Word: Danger