4-Ethylbenzaldehyde, 98%
4-Ethylbenzaldehyde, 98%
4-Ethylbenzaldehyde, 98%
4-Ethylbenzaldehyde, 98%
Thermo Scientific Chemicals
4-Ethylbenzaldehyde, 98%
CAS: 4748-78-1 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA378951000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Ethylbenzaldehyde
CAS4748-78-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0