tert-Butyl methyl ether, 99.9%, extra pure
tert-Butyl methyl ether, 99.9%, extra pure
tert-Butyl methyl ether, 99.9%, extra pure
tert-Butyl methyl ether, 99.9%, extra pure
Thermo Scientific Chemicals
tert-Butyl methyl ether, 99.9%, extra pure
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA378720025
Price (MYR)
722.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
722.00
BOT
Add to cart
Specifications
Chemical Name or Materialtert-Butyl methyl ether
Melting Point-110.0°C
CAS1634-04-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.