Titanium(IV) chloride, 1M solution in toluene, AcroSeal™
CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol
Have Questions?
Change view
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number FSA378168000
Price (MYR)
2,365.00
ML
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,365.00
ML
Add to cart
Chemical Identifiers
CAS7550-45-0, 108-88-3
Specifications
Chemical Name or MaterialTitanium(IV) chloride
Name Note1M Solution in Toluene
CAS7550-45-0
ColorOrange
Health Hazard 1GHS Signal Word: Danger