9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%
9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%
9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%
9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%
Thermo Scientific Chemicals
9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%
CAS: 161265-03-8 | C39H32OP2 | 578.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA378060250
Price (MYR)
5,221.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,221.00
EA
Add to cart
Specifications
Chemical Name or Material9, 9-Dimethyl-4, 5-bis(diphenylphosphino)xanthene
CAS161265-03-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0